Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 1 Antibody (F18), Novus Biologicals™
SDP

Catalog No. NB10064387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064387 0.1 mg
NB164387SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064387 Supplier Novus Biologicals Supplier No. NB10064387
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 3 publications

IFN-alpha D Monoclonal specifically detects IFN-alpha D in Mouse samples. It is validated for ELISA, Functional, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IFN-alpha D
Applications ELISA, Functional Assay, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone F18
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Blocking/Neutralizing 6.25 x 10^4 neatralizing units/mg
Gene Accession No. P01572
Gene Alias IFL, IFN, IFNA13, IFNAalphaIFN-alpha-1/13, IFN-ALPHA, IFN-alpha 1b, IFN-alphaD, interferon alpha 1b, interferon alpha-1/13, Interferon alpha-D, interferon, alpha 1, LeIF D, MGC138207, MGC138505, MGC138507
Gene Symbols IFNA1
Host Species Rat
Immunogen Made to Mouse IFN ALPHA
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3439
Test Specificity NB100-64387 recognizes most isoforms of natural and recombinant murine alpha interferon. It does not cross-react with murine beta or gamma interferon, nor with human alpha interferon.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.