Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 2 Antibody (N39), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP24398A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24398A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24398A30 Supplier Novus Biologicals Supplier No. NBP254398AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha 2 Monoclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IFN-alpha 2
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone N39
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Gene Symbols IFNA2
Host Species Mouse
Immunogen Purified recombinant human IFN-alpha 2 (Uniprot: P01563)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3440.0
Test Specificity Recognizes a protein of 16-27kDa, identified as human interferon-II) (IFN-(II). Its epitope maps between aa112-148 of IFN-II) (total aa172). This monoclonal antibody is specific for IFN-(II) and does not cross-react with IFN-(I). The site recognized by this monoclonal antibody is called site I and is responsible for the antiviral and anti-proliferative activities of IFN-(II). Epitopes of N27 and N39 monoclonal antibodys are different and represent a good combination of antibodies to set up an ELISA assay for the quantitation of IFN-(II) after viral infections. The IFN- family consists of 24 or more genes or pseudo-genes. IFN-II) is one of the two distinct families (I and II) of human IFN-. The -interferon are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.