Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 2 Antibody (ST29), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB210804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB210804 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB210804 Supplier Novus Biologicals Supplier No. NB1002479AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for ELISA

Specifications

Antigen IFN-alpha 2
Applications ELISA
Classification Monoclonal
Clone ST29
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Host Species Mouse
Immunogen Human interferon (HuIFN) produced by Namalwa cells (HuIFN-aN)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3440
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.