Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 2 Antibody (ST29) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002479 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002479 Supplier Novus Biologicals Supplier No. NB1002479
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for ELISA, Functional Assay.

Specifications

Antigen IFN-alpha
Applications ELISA, Functional Assay
Classification Monoclonal
Clone ST29
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Functional
Gene Alias IFNA, IFNA2, IFNA2b, IFN-alphaA, INFA2, interferon alpha 2
Gene Symbols IFNA1
Host Species Mouse
Immunogen Human interferon (HuIFN) produced by Namalwa cells (HuIFN-aN)
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3440
Test Specificity Interferon alpha (ST29). Cross-reacts with Human. Not yet tested in other species.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.