Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB083039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB083039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB083039 Supplier Novus Biologicals Supplier No. NBP299055R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFN-alpha 4 Polyclonal specifically detects IFN-alpha 4 in Human samples. It is validated for ELISA.

Specifications

Antigen IFN-alpha 4
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias IFN-alpha-4, IFN-alpha4a, INFA4, interferon alpha-4, Interferon alpha-4B, Interferon alpha-76, Interferon alpha-M1, interferon, alpha 4, MGC142200
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IFN-alpha 4 (NP_066546.1; Met1-Asp189).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3441
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.