Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha B2/IFN-alpha 8 Rabbit anti-Human, Clone: 3, Novus Biologicals™
SDP

Catalog No. NB020363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020363 100 μg
NB020364 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020363 Supplier Novus Biologicals Supplier No. NBP289440100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IFN-alpha B2/IFN-alpha 8 Monoclonal specifically detects IFN-alpha B2/IFN-alpha 8 in Human samples. It is validated for ELISA.

Specifications

Antigen IFN-alpha B2/IFN-alpha 8
Applications ELISA
Classification Monoclonal
Clone 3
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B′, Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B
Host Species Rabbit
Immunogen Recombinant Human IFN-alpha B2/IFN-alpha 8 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3445
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.