Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha/beta R1 Antibody (85228), HRP, Novus Biologicals™
SDP

Catalog No. NB365058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365058 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365058 Supplier Novus Biologicals Supplier No. FAB245H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen IFN-alpha/beta R1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 85228
Conjugate HRP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha/beta R1, Lys28-Lys436, Accession # AAA52730
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3454
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.