Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha/beta R1 Antibody (MM0336-8H64), PerCP, Novus Biologicals™
SDP

Catalog No. NBP211731P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211731P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211731P Supplier Novus Biologicals Supplier No. NBP211731PCP

Mouse Monoclonal Antibody

IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IFN-alpha/beta R1
Applications Flow Cytometry
Classification Monoclonal
Clone MM0336-8H64
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. AAA52730
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Gene Symbols IFNAR1
Host Species Mouse
Immunogen Human recombinant IFN-alpha/beta R1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3454
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.