Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha/beta R1 Rabbit anti-Mouse, Clone: 110, Novus Biologicals™
SDP

Catalog No. NB024947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024947 100 μg
NB024948 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024947 Supplier Novus Biologicals Supplier No. NBP290600100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IFN-alpha/beta R1 Monoclonal specifically detects IFN-alpha/beta R1 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IFN-alpha/beta R1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 110
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Host Species Rabbit
Immunogen Recombinant Mouse IFN-alpha/beta R1 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3454
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.