Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha/beta R2 Antibody (RM0251-5L16), FITC, Novus Biologicals™
SDP

Catalog No. NBP211743F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211743F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211743F Supplier Novus Biologicals Supplier No. NBP211743F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IFN-alpha/beta R2 Monoclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IFN-alpha/beta R2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0251-5L16
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2
Gene Symbols IFNAR2
Host Species Rat
Immunogen Mouse recombinant IFN-alpha/beta R2 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.