Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha H2/IFNA14 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB132179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132179 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132179 Supplier Novus Biologicals Supplier No. NBP306094DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFN-alpha H2/IFNA14 Polyclonal specifically detects IFN-alpha H2/IFNA14 in Mouse samples. It is validated for ELISA.

Specifications

Antigen IFN-alpha H2/IFNA14
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias IFN-alpha-14, IFN-alphaH, interferon alpha-14, Interferon alpha-H, Interferon lambda-2-H, interferon, alpha 14, LeIF H, LEIF2H, MGC125756, MGC125757
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse IFN-alpha H2/IFNA14 (NP_996858.1; Cys24-Lys189).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3448
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.