Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha J1/IFNA7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
| Antigen | IFN-alpha J1/IFNA7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179586
![]() |
Novus Biologicals
NBP179586 |
100 μL | N/A | N/A | N/A | ||||
Description
IFN-alpha J1/IFNA7 Polyclonal specifically detects IFN-alpha J1/IFNA7 in Human samples. It is validated for Western Blot.Specifications
| IFN-alpha J1/IFNA7 | |
| Polyclonal | |
| Rabbit | |
| NP_066401 | |
| 3444 | |
| Synthetic peptide directed towards the N terminal of human IFNA7The immunogen for this antibody is IFNA7. Peptide sequence RALILLAQMGRISPFSCLKDRHEFRFPEEEFDGHQFQKTQAISVLHEMIQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| interferon alpha-7, interferon alpha-J1, interferon, alpha 7 | |
| IFNA7 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title