Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha K Antibody (OTI2A10), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271007B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271007B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271007B Supplier Novus Biologicals Supplier No. NBP271007B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-alpha K Monoclonal specifically detects IFN-alpha K in Human samples. It is validated for Western Blot.

Specifications

Antigen IFN-alpha K
Applications Western Blot
Classification Monoclonal
Clone OTI2A10
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias IFNA6, IFN-alpha 6, IFNalpha K, IFN-alpha K, IFN-kappa, interferon kappa, interferon, kappa, RP11-27J8.1, UNQ6124/PRO20084
Gene Symbols IFNA6
Host Species Mouse
Immunogen Full length recombinant protein of human IFN-alpha K (NP_066282) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3443.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.