Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (154007), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB302288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302288 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302288 Supplier Novus Biologicals Supplier No. FAB985R

Mouse Monoclonal Antibody

IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Porcine samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IFN-gamma
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 154007
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IFN-gamma , Ser21-Lys166, Accession # P17803
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.