Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (25723), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348693 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348693 Supplier Novus Biologicals Supplier No. FAB2851MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Neutralization.

Specifications

Antigen IFN-gamma
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Neutralization
Classification Monoclonal
Clone 25723
Conjugate mFluor Violet 450 SE
Dilution Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-gamma , Gln24-Gln166, Accession # AAP20098.1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.