Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (37895R), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB368910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368910 Supplier Novus Biologicals Supplier No. FAB485RE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization.

Specifications

Antigen IFN-gamma
Applications Flow Cytometry (Intracellular Staining), Neutralization
Classification Monoclonal
Clone 37895R
Conjugate DyLight 405
Dilution Neutralization, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Rat
Immunogen E. coli-derived recombinant mouse IFN-gamma
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.