Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (D9D10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB014728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014728 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014728 Supplier Novus Biologicals Supplier No. NBP281044AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen IFN-gamma
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone D9D10
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Rabbit
Immunogen FPLC-purified natural human IFN-gamma. Refer to the original article [PMID: 3114127]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Binds human IFN-gamma, a dimerized soluble cytokine that is the only member of the type II class of interferons.
Target Species Human, Baboon
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.