Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (N2P2D5-A10), Novus Biologicals™
SDP

Catalog No. NB010671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010671 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010671 Supplier Novus Biologicals Supplier No. NBP279821

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Fish, Salmon samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IFN-gamma
Applications Western Blot, ELISA
Classification Monoclonal
Clone N2P2D5-A10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Mouse
Immunogen Synthetic peptide from the mid-protein of Salmonid IFN gamma
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Target Species Human, Mouse, Fish
Content And Storage Store at -20 °C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.