Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Antibody (N3-P3A5*A10), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP2043A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2043A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2043A70 Supplier Novus Biologicals Supplier No. NBP250435AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IFN-gamma
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone N3-P3A5*A10
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.