Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R1/CD119 Antibody (062), HRP, Novus Biologicals™
SDP

Catalog No. NB316257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316257 Supplier Novus Biologicals Supplier No. NBP290685H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IFN-gamma R1/CD119 Monoclonal antibody specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, ELISA

Specifications

Antigen IFN-gamma R1/CD119
Applications Flow Cytometry, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 062
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA Capture
Formulation PBS
Gene Alias AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN-gamma R1/CD119 (Uniprot#: P15261; Met1-Asp253).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3459
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.