Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R1/CD119 Antibody (92101), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311529 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311529 Supplier Novus Biologicals Supplier No. FAB6731PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-gamma R1/CD119 Monoclonal antibody specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IFN-gamma R1/CD119
Applications Flow Cytometry
Classification Monoclonal
Clone 92101
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1, Glu18-Gly245, Accession # P15260.1
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3459
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.