Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R1/CD119 Rabbit anti-Mouse, Clone: 74, Novus Biologicals™
SDP

Catalog No. NB025120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025120 50 μg
NB025119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025120 Supplier Novus Biologicals Supplier No. NBP29068650UL

Rabbit Monoclonal Antibody

IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen IFN-gamma R1/CD119
Applications ELISA
Classification Monoclonal
Clone 74
Conjugate Unconjugated
Dilution Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Host Species Rabbit
Immunogen Recombinant Mouse IFN-gamma R1/CD119 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3459
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.