Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Antibody (MOB47), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311146 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311146 Supplier Novus Biologicals Supplier No. FAB773PECY55
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IFN-gamma R2
Applications Flow Cytometry
Classification Monoclonal
Clone MOB47
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Host Species Hamster
Immunogen Recombinant mouse IFN-gamma R2, Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3460
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.