Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Antibody (OTI1C2), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271008A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271008A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271008A700 Supplier Novus Biologicals Supplier No. NBP271008AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IFN-gamma R2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C2
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Gene Symbols IFNGR2
Host Species Mouse
Immunogen Full length recombinant protein of human IFN-gamma R2 (NP_005525) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3460.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.