Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma R2 Antibody (RM0252-7A25) - Azide and BSA Free, Novus Biologicals™

Hamster Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | IFN-gamma R2 |
|---|---|
| Clone | RM0252-7A25 |
| Concentration | LYOPH |
| Applications | CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Blocking Assay, Western Blot |
| Classification | Monoclonal |
Description
IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready, Block/Neutralize.Specifications
| IFN-gamma R2 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| Mouse | |
| AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
| IFNGR2 | |
| Primary |
| RM0252-7A25 | |
| CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Blocking Assay, Western Blot | |
| Unconjugated | |
| RUO | |
| AAH55745.1 | |
| 3460 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title