Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Antibody (RM0252-7A25) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019214 0.025 mg
NBP212430 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019214 Supplier Novus Biologicals Supplier No. NBP2124300.025MG
Only null left
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready, Block/Neutralize.

Specifications

Antigen IFN-gamma R2
Applications CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Blocking Assay, Western Blot
Classification Monoclonal
Clone RM0252-7A25
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. AAH55745.1
Gene Alias AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Gene Symbols IFNGR2
Host Species Hamster
Immunogen Mouse recombinant protein of IFNGR2, extracellular domain
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3460
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.