Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Antibody (RM0252-7A25), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212430UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212430UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212430UV Supplier Novus Biologicals Supplier No. NBP212430UV
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

IFN-gamma R2 Monoclonal antibody specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IFN-gamma R2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone RM0252-7A25
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Gene Alias AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Gene Symbols IFNGR2
Host Species Armenian Hamster
Immunogen Mouse recombinant IFN-gamma R2 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3460
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.