Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN Induced GTPase-like Protein Antibody, Novus Biologicals™
SDP

Catalog No. NBP244169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244169 Supplier Novus Biologicals Supplier No. NBP244169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects IFN Induced GTPase-like Protein in Mouse samples, and it is validated for ELISA, Western Blotting

IFN Induced GTPase-like Protein Polyclonal specifically detects IFN Induced GTPase-like Protein in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IFN Induced GTPase-like Protein
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 86.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:2000-1:10000
Formulation 0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. E9QAA8
Gene Alias EG225594, Ifgga3, interferon-gamma induced, Interferon-gamma-inducible GTPase Ifgga3 protein
Gene Symbols GM4841
Host Species Rabbit
Immunogen IFN Induced GTPase-like Protein Protein was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-terminal region of mouse IFN Induced GTPase-like Protein Protein. (Uniprot: E9QAA8)
Molecular Weight of Antigen 47.82 kDa
Purification Method Delipidation and Defibrination
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.