Learn More
IFN-γ Mouse anti-Human, APC, Clone: B27, BD
Mouse Monoclonal Antibody
$132.60 - $360.00
Specifications
Antigen | IFN-γ |
---|---|
Clone | B27 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB562017
|
BD Biosciences
562017 |
25 μg |
Each for $132.60
|
|
|||||
BDB554702
|
BD Biosciences
554702 |
0.1 mg |
Each for $360.00
|
|
|||||
Description
The B27 monoclonal antibody specifically binds to human interferon- γ(IFN- γ). IFN-γis a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ + T cells. IFN- γexerts i ts biological effects through specific binding to the high-affinity IFN- γReceptor Complex comprised of IFN- γβ subunits. In addition to its antiviral effects, IFN- γupregulates a number of lymphoid cell functions including the antimicrobial and antitumor responses of macrophages, NK cells, and neutrophils. In addition, IFN- γcan exert strong regulatory influences on the proliferation, differentiation, and effector responses of B cell and T cell subsets. Th ese influences can involve IFN- γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells t hemselves. Human IFN-γis a 14- 18 kDa glycoprotein containing 143 amino acid residues. B27 is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN- γhas been described. The B27 antibody has been reported not to bind to denatured IFN- γ.
Specifications
IFN-γ | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
IFNG; Interferon-gamma; Interferon-γ; Type II interferon; MAF |
B27 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.