Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-γ, Mouse Anti-Human, RY610, B27, BD Horizon™
Mouse Monoclonal Antibody
$176.00 - $406.00
Specifications
Antigen | IFN-γ |
---|---|
Clone | B27 |
Applications | Flow Cytometry (Intracellular Staining) |
Classification | Monoclonal |
Conjugate | RY610 |
Description
IFN-γ Monoclonal antibody specifically detects IFN-γ in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
IFN-γ | |
Flow Cytometry (Intracellular Staining) | |
RY610 | |
Mouse | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
IFNG; Interferon-gamma; ; Type II interferon; MAF | |
Human IFN-γRecombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B27 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3458 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title