Learn More
Invitrogen™ IFNAR1 Monoclonal Antibody (MAR1-5A3), PE, eBioscience™

Mouse Monoclonal Antibody
$382.00
Specifications
| Antigen | IFNAR1 |
|---|---|
| Clone | MAR1-5A3 |
| Concentration | 0.2 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-2359
![]()
|
Invitrogen™
12594582 |
100 μg |
Each of 1 for $382.00
|
|
|||||
Description
Description: The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities. Applications Reported: This MAR1-5A3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MAR1-5A3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).Specifications
| IFNAR1 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P33896 | |
| 15975 | |
| Primary | |
| Affinity chromatography |
| MAR1-5A3 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Mouse | |
| Ifnar1 | |
| alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1 | |
| Ifnar1 | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.