Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFRD1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB324534 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324534 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324534 Supplier Novus Biologicals Supplier No. NBP299593PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFRD1 Polyclonal antibody specifically detects IFRD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IFRD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias 12-O-tetradecanoylphorbol-13-acetate-induced sequence 7, interferon-related developmental regulator 1, Nerve growth factor-inducible protein PC4, PC4, pheochromocytoma cell-4, TIS7, TPA induced sequence 7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IFRD1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3475
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.