Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT52 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731630 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1630 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1630 Supplier Proteintech Group Inc Supplier No. 175341AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintece and functioning of primary cilia.

Specifications

Antigen IFT52
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFT52
Gene Accession No. Q62559, Q9Y366
Gene Alias C20orf9, CGI 53, IFT52, NGD5, Protein NGD5 homolog
Gene Symbols IFT52
Host Species Rabbit
Immunogen IFT52 Fusion Protein Ag11586
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245866, 362265, 51098, 610842
Target Species Canine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.