Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFT52 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | IFT52 |
|---|---|
| Concentration | 0.38 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1630
![]()
|
Proteintech Group Inc
175341AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintece and functioning of primary cilia.Specifications
| IFT52 | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Canine, Human, Mouse, Rat | |
| Q62559, Q9Y366 | |
| 245866, 362265, 51098, 610842 | |
| IFT52 Fusion Protein Ag11586 | |
| Primary | |
| -20°C | |
| IFT52 |
| 0.38 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C20orf9, CGI 53, IFT52, NGD5, Protein NGD5 homolog | |
| IFT52 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title