Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT57 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$231.00 - $516.29

Specifications

Antigen IFT57
Clone OTI2G3
Concentration 0.71 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-254-2688
SDP
Encompass
Encompass_Preferred
View Documents
OriGene
TA500917S
30 μL
Each for $231.00
Only null left
Add to Cart
 
50-167-2058
SDP
Encompass_Preferred
View Documents
OriGene
TA500917
100 μL
Each for $516.29
Only null left
Add to Cart
 
Description

Description

IFT57 Monoclonal antibody specifically detects IFT57 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

IFT57
0.71 mg/mL
Unconjugated
Mouse
Human
Primary
OTI2G3
Monoclonal
Liquid
RUO
IgG1
-20° C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.