Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ift80 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174179
Description
Ift80 Polyclonal specifically detects Ift80 in Mouse samples. It is validated for Western Blot.Specifications
Ift80 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ATD2, intraflagellar transport 80 homolog (Chlamydomonas), intraflagellar transport protein 80 homolog, KIAA1374WDR56WD repeat domain 56, MGC126543, WD repeat-containing protein 56 | |
Rabbit | |
85 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Equine: 92%; Rabbit: 92%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8K057 | |
IFT80 | |
Synthetic peptides corresponding to the C terminal of Ift80. Immunizing peptide sequence PNTIYVDRDILPKTLYERDASEYSKNPHIVSFVGNQVTIRRADGSLVHIS. | |
Affinity purified | |
RUO | |
57560 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction