Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ift80 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Ift80
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP174179
SDP
View Documents
Novus Biologicals
NBP174179
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Ift80 Polyclonal specifically detects Ift80 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Ift80
Polyclonal
Rabbit
Q8K057
57560
Synthetic peptides corresponding to the C terminal of Ift80. Immunizing peptide sequence PNTIYVDRDILPKTLYERDASEYSKNPHIVSFVGNQVTIRRADGSLVHIS.
Primary
Western Blot
Unconjugated
RUO
ATD2, intraflagellar transport 80 homolog (Chlamydomonas), intraflagellar transport protein 80 homolog, KIAA1374WDR56WD repeat domain 56, MGC126543, WD repeat-containing protein 56
IFT80
IgG
85 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.