Learn More
IFT88 Mouse anti-Human, Mouse, Rat, Clone: 4A4G5, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 602271IG150UL

Description
This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
IFT88 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13099, Q61371 | |
IFT88 | |
IFT88 Fusion Protein Ag4980 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
4A4G5 | |
Unconjugated | |
IFT88 | |
IFT88, TPR repeat protein 10, TTC10 | |
Mouse | |
Protein G | |
RUO | |
21821, 305918, 8100 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.