Learn More
IFT88 Mouse anti-Human, Mouse, Rat, Clone: 4A4G5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IFT88 |
---|---|
Clone | 4A4G5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6328
![]() ![]() |
Proteintech Group Inc
602271IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
IFT88 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IFT88, TPR repeat protein 10, TTC10 | |
IFT88 | |
IgG1 | |
Protein G | |
Antibody |
4A4G5 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q13099, Q61371 | |
21821, 305918, 8100 | |
IFT88 Fusion Protein Ag4980 | |
Primary | |
-20°C | |
IFT88 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.