Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig gamma-4 Chain C Region Antibody (IGHG4/1345), Novus Biologicals™
SDP

Catalog No. NB253218200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253218200 200 μg
NB25321820 20 μg
NB253218100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253218200 Supplier Novus Biologicals Supplier No. NBP253218200UG

Mouse Monoclonal Antibody

Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Ig gamma-4 Chain C Region
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IGHG4/1345
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker)
Host Species Mouse
Immunogen Recombinant human full-length IGHG4
Molecular Weight of Antigen 150 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3503
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.