Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig gamma-4 Chain C Region Antibody (IGHG4/2042R), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB011059 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011059 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011059 Supplier Novus Biologicals Supplier No. NBP279850FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ig gamma-4 Chain C Region
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IGHG4/2042R
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker)
Gene Symbols IGHG4
Host Species Rabbit
Immunogen Recombinant full-length human Ig gamma-4 Chain C Region protein (Uniprot: P01861)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.