Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig gamma-4 Chain C Region Antibody (rIGHG4/1345), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB385204 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385204 Supplier Novus Biologicals Supplier No. NBP308668AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Ig gamma-4 Chain C Region
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rIGHG4/1345
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker)
Host Species Mouse
Immunogen Recombinant human Ig gamma-4 Chain C Region fragment (Uniprot: P01861)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3503
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.