Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig Light Chain Antibody (RM129) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301082 Supplier Novus Biologicals Supplier No. NBP325967
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ig Light Chain Monoclonal antibody specifically detects Ig Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Ig Light Chain
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone RM129
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5ug/ml-2ug/ml, ELISA : 0.1 ug/ml - 0.5 ug/ml, Immunohistochemistry : 0.5ug/ml - 2ug/ml, Immunocytochemistry/ Immunofluorescence : 0.5ug/ml - 2ug/ml
Formulation PBS
Host Species Rabbit
Immunogen Human IgG
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.