Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Antibody (RM128) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301080 Supplier Novus Biologicals Supplier No. NBP325969
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgA Monoclonal antibody specifically detects IgA in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ELISA, ELISA

Specifications

Antigen IgA
Applications ELISA, Immunohistochemistry, Immunocytochemistry, ELISA, ELISA
Classification Monoclonal
Clone RM128
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry : 0.1 - 1ug/ml, Immunocytochemistry/ Immunofluorescence : 0.5 - 2ug/ml, Sandwich ELISA Capture : 50 - 200ng/well, Sandwich ELISA Detection : 0.05 - 0.2ug/ml
Formulation PBS
Gene Alias Immunoglobulin A
Host Species Rabbit
Immunogen Human IgA
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3493
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.