Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Mouse anti-Human, Alexa Fluor™ 750, Clone: AD3, Novus Biologicals™
SDP

Catalog No. NB50469A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50469A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50469A750 Supplier Novus Biologicals Supplier No. NB500469AF750

Mouse Monoclonal Antibody

IgA Monoclonal specifically detects IgA in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IgA
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AD3
Conjugate Alexa Fluor 750
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Mouse
Immunogen Purified human IgA.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3493
Test Specificity The antibody AD3 reacts with alpha-chain of human IgA1 and IgA2.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.