Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Secretory Chain Antibody (Mc29-12), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N106476A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106476A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106476A32 Supplier Novus Biologicals Supplier No. NB10064756AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgA Secretory Chain Monoclonal specifically detects IgA Secretory Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen IgA Secretory Chain
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone Mc29-12
Conjugate Alexa Fluor 532
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686C15213, DKFZp686K04218, DKFZp686L19235, DKFZp686M08189, FLJ41981, FLJ43089, FLJ44381, IGH, IGH.1alpha, IGHDY1, immunglobulin heavy chain variable region, immunoglobulin heavy locus, MGC70893, MGC72071, MGC88774
Gene Symbols IGH
Host Species Mouse
Immunogen Made to Human IgA
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3492
Test Specificity NB100-64756 reacts strongly to purified secretory - piece and secretory IgA. No reaction against myeloma IgA (native or reduced and alkylated). Negative against normal human IgM, myeloma IgM, normal human IgG; free kappa or gamma chains, and J-chain. Typical pattern on frozen and paraffin embedded sections of intestinal and breast tissue.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.