Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Secretory Chain Antibody (Mc29-12), Novus Biologicals™
SDP

Catalog No. NB017636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017636 0.025 mg
NB10064756 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017636 Supplier Novus Biologicals Supplier No. NB100647560.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgA Secretory Chain Monoclonal specifically detects IgA Secretory Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen IgA Secretory Chain
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Mc29-12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation TBS with 0.09% Sodium Azide
Gene Accession No. P01876
Gene Alias DKFZp686C15213, DKFZp686K04218, DKFZp686L19235, DKFZp686M08189, FLJ41981, FLJ43089, FLJ44381, IGH, IGH.1alpha, IGHDY1, immunglobulin heavy chain variable region, immunoglobulin heavy locus, MGC70893, MGC72071, MGC88774
Gene Symbols IGH
Host Species Mouse
Immunogen Made to Human IgA
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3492
Test Specificity NB100-64756 reacts strongly to purified secretory - piece and secretory IgA. No reaction against myeloma IgA (native or reduced and alkylated). Negative against normal human IgM, myeloma IgM, normal human IgG; free kappa or gamma chains, and J-chain. Typical pattern on frozen and paraffin embedded sections of intestinal and breast tissue.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.