Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA1 Antibody (RM124) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301079 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301079 Supplier Novus Biologicals Supplier No. NBP325970
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgA1 Monoclonal antibody specifically detects IgA1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ELISA, ELISA

Specifications

Antigen IgA1
Applications ELISA, Immunohistochemistry, Immunocytochemistry, ELISA, ELISA
Classification Monoclonal
Clone RM124
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry : 0.1 - 1ug/ml, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA Capture : 50 - 200ng/well, Sandwich ELISA Detection : 0.05 - 0.2ug/ml
Formulation PBS
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Host Species Rabbit
Immunogen Human IgA
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3493
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.