Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (11-26c.2a), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP149000AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149000AH 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149000AH Supplier Novus Biologicals Supplier No. NBP149000AF647

This product is either new or inactive, and cannot be ordered online.

Rat Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgD
Applications Flow Cytometry
Classification Monoclonal
Clone 11-26c.2a
Conjugate Alexa Fluor 647
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Gene Symbols IGHD
Host Species Rat
Immunogen This antibody was generated by immunizing host with mouse IgD
Purification Method Protein A or G purified
Quantity 0.5 mL
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.