Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (IgD26), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB379569 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379569 Supplier Novus Biologicals Supplier No. NBP308605V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen IgD
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IgD26
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Host Species Mouse
Immunogen Full-length human IgD protein (Uniprot: P01880)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.